Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USH1C Monoclonal Antibody (2B3)
GREENER_CHOICE

Catalog No. PIMA520917
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA520917 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA520917 Supplier Invitrogen™ Supplier No. MA520917
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: FTPEQIMGKD VRLLRIKKEG SLDLALEGGV DSPIGKVVVS AVYERGAAER HGGIVKGDEI MAINGKIVTD YTLAEADAAL QKAWNQGGDW IDLVVAVCPP KEYDDELTFF.

This gene encodes a scaffold protein that functions in the assembly of Usher protein complexes. The protein contains PDZ domains, a coiled-coil region with a bipartite nuclear localization signal and a PEST degradation sequence. Defects in this gene are the cause of Usher syndrome type 1C and non-syndromic sensorineural deafness autosomal recessive type 18. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USH1C
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2B3
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene USH1C
Gene Accession No. Q9Y6N9
Gene Alias 2010016F01Rik; AIE75; AIE-75; Antigen NY-CO-38/NY-CO-37; autoimmune enteropathy-related antigen AIE-75; DFNB18; DFNB18A; harmonin; harmonin a1; harmonin pseudogene; hypothetical protein LOC530709; hypothetical protein LOC564412; NY-CO-37; NY-CO-38; PDZ domain-containing protein; PDZ-45; PDZ73; PDZ-73; PDZ-73 protein; PDZ-73/NY-CO-38; PDZD7C; Protein PDZ-73; Renal carcinoma antigen NY-REN-3; USH1 protein network component harmonin; Ush1c; ush1cpst; Usher syndrome 1C; Usher syndrome 1C (autosomal recessive, severe); Usher syndrome 1C homolog; usher syndrome type-1C protein; Usher syndrome type-1C protein homolog; zgc:136806
Gene Symbols USH1C
Host Species Mouse
Immunogen USH1C (AAH16057, 424 a.a. ~ 533 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10083
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.