Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USH1C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588622
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588622 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588622 Supplier Invitrogen™ Supplier No. PA588622
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VDFSNLDHKE GRELFMTDRE RLAEARQREL QRQELLMQKR LAMESNKILQ EQQEMERQRR KEIAQKAAEE NERYRKEMEQ IVEEEEKFKK QWEEDWGSKE QLLLPKTITA EVHPVPLRKP KYDQGVEPEL EPADDLDGGT EEQGEQDFRK YEEGFDPYSM FTPEQIMGKD VRLLRIKKEG SLDLALEGGV DSPIGKVVVS AVYERGAAER HGGIVKGDEI MAINGKIVTD YTLAEAEAAL QKAWNQGGDW IDLVVAVCPP KEYDDELTFF; Positive Samples: 293T, HeLa, Mouse kidney, Mouse intestine, Rat kidney; Cellular Location: Cytoplasm, cytoskeleton, cytosol.

This gene encodes a scaffold protein that functions in the assembly of Usher protein complexes. The protein contains PDZ domains, a coiled-coil region with a bipartite nuclear localization signal and a PEST degradation sequence. Defects in this gene are the cause of Usher syndrome type 1C and non-syndromic sensorineural deafness autosomal recessive type 18. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USH1C
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.83 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene USH1C
Gene Accession No. Q9ES64, Q9Y6N9
Gene Alias 2010016F01Rik; AIE75; AIE-75; Antigen NY-CO-38/NY-CO-37; autoimmune enteropathy-related antigen AIE-75; DFNB18; DFNB18A; harmonin; harmonin a1; harmonin pseudogene; hypothetical protein LOC530709; hypothetical protein LOC564412; NY-CO-37; NY-CO-38; PDZ domain-containing protein; PDZ-45; PDZ73; PDZ-73; PDZ-73 protein; PDZ-73/NY-CO-38; PDZD7C; Protein PDZ-73; Renal carcinoma antigen NY-REN-3; USH1 protein network component harmonin; Ush1c; ush1cpst; Usher syndrome 1C; Usher syndrome 1C (autosomal recessive, severe); Usher syndrome 1C homolog; usher syndrome type-1C protein; Usher syndrome type-1C protein homolog; zgc:136806
Gene Symbols USH1C
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 264-533 of human USH1C (NP_0012846931).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10083, 308596, 72088
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.