Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ USH1C Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116999
Description
Positive Samples: HT-29, Mouse kidney, Mouse small intestine, Rat testis, Rat kidney Immunogen sequence: MERKVAREFR HKVELLIDNE AEKDYLYDVL RMYHQSMDLP VLVGDLKLVI NEPKRLPLFD AIRPLIPLKH QVQYDQLTPK RSRKLKEVRL DRTHPEGLGL.
This gene encodes a scaffold protein that functions in the assembly of Usher protein complexes. The protein contains PDZ domains, a coiled-coil region with a bipartite nuclear localization signal and a PEST degradation sequence. Defects in this gene are the cause of Usher syndrome type 1C and non-syndromic sensorineural deafness autosomal recessive type 18. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| USH1C | |
| Polyclonal | |
| Unconjugated | |
| USH1C | |
| 2010016F01Rik; AIE75; AIE-75; Antigen NY-CO-38/NY-CO-37; autoimmune enteropathy-related antigen AIE-75; DFNB18; DFNB18A; harmonin; harmonin a1; harmonin pseudogene; hypothetical protein LOC530709; hypothetical protein LOC564412; NY-CO-37; NY-CO-38; PDZ domain-containing protein; PDZ-45; PDZ73; PDZ-73; PDZ-73 protein; PDZ-73/NY-CO-38; PDZD7C; Protein PDZ-73; Renal carcinoma antigen NY-REN-3; USH1 protein network component harmonin; Ush1c; ush1cpst; Usher syndrome 1C; Usher syndrome 1C (autosomal recessive, severe); Usher syndrome 1C homolog; usher syndrome type-1C protein; Usher syndrome type-1C protein homolog; zgc:136806 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10083, 308596, 72088 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9ES64, Q9Y6N9 | |
| USH1C | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of zebrafish USH1C (NP_0010350181). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction