Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USHBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100278
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100278 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100278 Supplier Invitrogen™ Supplier No. PA5100278
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MCC.

USHBP1, also named as Mutated in colon cancer protein 2 or AIE-75-binding protein, is a 703 amino acid protein, which belongs to the MCC family. Highest level of expression is in heart, and moderate to low expression in skeletal muscle, kidney, liver, small intestine, placenta and lung.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USHBP1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene USHBP1
Gene Accession No. Q8N6Y0, Q8R370
Gene Alias 2210404N08Rik; 9430064H24; AIE-75 binding protein; AIE-75-binding protein; AIEBP; MCC2; MCC-2; mutated in colon cancer protein 2; USH1 protein network component harmonin binding protein 1; USH1C-binding protein 1; USHBP1; Usher syndrome 1C binding protein 1; Usher syndrome type-1C protein-binding protein 1
Gene Symbols USHBP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human USHBP1(Accession Q8N6Y0), corresponding to amino acid residues L389-M439.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 234395, 83878
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.