Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USHBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562618
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562618 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562618 Supplier Invitrogen™ Supplier No. PA562618
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TLQHTLSSLE AAAAAWRHQP PSHSGPMEFE GTSEGGAGSL GKQEGAGSCQ REAARLAERN AWLRLALSSR EDELVRTQAS LEAIRAEKET LQKEVQELQD SLLRLEPCPH LSHNQAGG Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 56%.

USHBP1, also named as Mutated in colon cancer protein 2 or AIE-75-binding protein, is a 703 amino acid protein, which belongs to the MCC family. Highest level of expression is in heart, and moderate to low expression in skeletal muscle, kidney, liver, small intestine, placenta and lung.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USHBP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene USHBP1
Gene Accession No. Q8N6Y0
Gene Alias 2210404N08Rik; 9430064H24; AIE-75 binding protein; AIE-75-binding protein; AIEBP; MCC2; MCC-2; mutated in colon cancer protein 2; USH1 protein network component harmonin binding protein 1; USH1C-binding protein 1; USHBP1; Usher syndrome 1C binding protein 1; Usher syndrome type-1C protein-binding protein 1
Gene Symbols USHBP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human USHBP1. Recombinant protein control fragment (Product #RP-99073).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83878
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.