Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Usherin Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144075
Description
Usherin Polyclonal Antibody for IHC (F)
Involved in hearing and vision as member of the USH2 complex. In the inner ear, required for the maintenance of the hair bundle ankle formation, which connects growing stereocilia in developing cochlear hair cells. In retina photoreceptors, the USH2 complex is required for the maintenance of periciliary membrane complex that seems to play a role in regulating intracellular protein transport.
Specifications
Usherin | |
Polyclonal | |
Unconjugated | |
USH2A | |
USH2A; Usherin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
568982 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Frozen), Immunocytochemistry, Immunohistochemistry (Frozen) | |
500 μg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q5FVR0, Q8R183 | |
USH2A | |
Zebrafish USH2A between 5081-5236. | |
200 μL | |
Primary | |
Zebrafish | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction