Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Usherin Rabbit anti-Human, Mouse, Non-human Primate, Rat, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc USH101AP

Description
Usherin Polyclonal antibody specifically detects Usherin in Human, Mouse, Non-human Primate, Rat samples. It is validated for ELISA, ImmunoprecipitationSpecifications
Usherin | |
Polyclonal | |
Unconjugated | |
USH2A | |
USH2A; Usherin | |
Synthetic peptide taken within amino acid region 150-180 on usherin isoform A protein. | |
200 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
ELISA, Immunoprecipitation | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
O75445, Q2QI47, Q8K3K1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
22283, 289369, 7399 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction