Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USMG5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565390
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565390 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565390 Supplier Invitrogen™ Supplier No. PA565390
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAGPESDAQYQ FTGIKKYFNS YTLTG Highest antigen sequence identity to the following orthologs - mouse 88%, rat 92%.

Plays a critical role in maintaining the ATP synthase population in mitochondria. Target protein is a novel protein that is upregulated in streptozotocin-induced diabetic rats.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USMG5
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene USMG5
Gene Accession No. Q96IX5
Gene Alias 2010301L15Rik; AGP; ATP synthase membrane subunit DAPIT; ATP synthase membrane subunit DAPIT, mitochondrial; Atp5md; bA792D24.4; Dapit; Diabetes Associated Protein in Insulin-sensitive Tissues; diabetes-associated protein in insulin-sensitive tissue; diabetes-associated protein in insulin-sensitive tissues; HCV F-transactivated protein 2; HCVFTP2; PD04912; Unknown (protein for MGC:128614); upregulated during skeletal muscle growth 5; up-regulated during skeletal muscle growth 5; upregulated during skeletal muscle growth 5 homolog; up-regulated during skeletal muscle growth 5 homolog; up-regulated during skeletal muscle growth 5 homolog (mouse); up-regulated during skeletal muscle growth protein 5; Usmg5
Gene Symbols USMG5
Host Species Rabbit
Immunogen Recombinant Human USMG5. Recombinant protein control fragment (Product #RP-106072).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84833
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.