Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USMG5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569671
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569671 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569671 Supplier Thermo Scientific Supplier No. PA569671
Add to Cart
Add to Cart

USMG5 Polyclonal Antibody

USMG5 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen USMG5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene USMG5
Gene Accession No. Q96IX5
Gene Alias 2010301L15Rik; AGP; ATP synthase membrane subunit DAPIT; ATP synthase membrane subunit DAPIT, mitochondrial; Atp5md; bA792D24.4; Dapit; Diabetes Associated Protein in Insulin-sensitive Tissues; diabetes-associated protein in insulin-sensitive tissue; diabetes-associated protein in insulin-sensitive tissues; HCV F-transactivated protein 2; HCVFTP2; PD04912; Unknown (protein for MGC:128614); upregulated during skeletal muscle growth 5; up-regulated during skeletal muscle growth 5; upregulated during skeletal muscle growth 5 homolog; up-regulated during skeletal muscle growth 5 homolog; up-regulated during skeletal muscle growth 5 homolog (mouse); up-regulated during skeletal muscle growth protein 5; Usmg5
Gene Symbols USMG5
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human USMG5.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84833
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.