Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP 467 Calibration Mixture #3, Restek™
SDP

Catalog No. 06802561 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-802-561 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-802-561 Supplier Restek Corporation Supplier No. 36004
  • The Class 1 mixtures below reflect requirements of USP23/NF18 effective January 1, 1995 to December 31, 1999.
  • Mixtures do not meet the current USP guidelines, many still use these mixtures to obtain a detectable benzene peak for the direct injection methods, Method I and Method V

Compounds:
 

  • Benzene (71-43-2), 100 μg/mL
  • Chloroform (67-66-3), 50μg/mL
  • 1,4-Dioxane (123-91-1), 100μg/mL
  • Methylene chloride (dichloromethane) (75-09-2), 500μg/mL
  • Trichloroethene (79-01-6), 100μg/mL

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methanesulfinylmethane
SMILES CS(C)=O

Specifications

CAS Max % 99.150000
Odor Strong
Specific Gravity 1.104 g/cm3 at 20°C
Concentration In dimethyl sulfoxide, 1 mL/ampul
Melting Point 18.5°C
Type Multi-component Standard
Quantity 1 mL
Chemical Name or Material Dimethyl Sulfoxide
ShelfLife 6 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.