Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP1 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10088117DL594
Description
USP1 Polyclonal antibody specifically detects USP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
USP1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Deubiquitinating enzyme 1, EC 3.1.2.15, EC 3.4.19.12, hUBP, ubiquitin carboxyl terminal hydrolase 1, ubiquitin carboxyl-terminal hydrolase 1, ubiquitin specific peptidase 1, ubiquitin specific processing protease 1, ubiquitin specific protease 1, ubiquitin thioesterase 1, Ubiquitin thiolesterase 1, Ubiquitin-specific-processing protease 1, UBP | |
The immunogen recognized by this antibody maps to a region between residue 450 and 500 of human ubiquitin-specific-processing protease 1 using the numbering given in entry NP_003359.3 (GeneID 7398). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
7398 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction