Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP1 Mouse anti-Human, Clone: 1C10H6, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | USP1 |
---|---|
Clone | 1C10H6 |
Concentration | 1.37 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6506
![]() ![]() |
Proteintech Group Inc
660691IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Deubiquitinating enzymes (DUBs) are capable of producing a free ubiquitin moiety from ubiquitin-fused precursors and ubiquitinylated proteins in cells. DUBs possess conserved catalytic regions known as the ′cys domain′ and the ′his domain.′ There are 2 distinct families of DUBs: ubiquitin-specific processing proteases (UBPs, or USPs) and ubiquitin C-terminal hydrolases (UCHs).Specifications
USP1 | |
1.37 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Deubiquitinating enzyme 1, hUBP, ubiquitin specific peptidase 1, Ubiquitin thiolesterase 1, UBP, USP1 | |
USP1 | |
IgG2a | |
Protein A | |
Antibody |
1C10H6 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
O94782 | |
7398 | |
USP1 Fusion Protein Ag5904 | |
Primary | |
-20°C | |
USP1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title