Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119620
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119620 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119620 Supplier Invitrogen™ Supplier No. PA5119620
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Deubiquitinating enzymes (DUBs) are capable of producing a free ubiquitin moiety from ubiquitin-fused precursors and ubiquitinylated proteins in cells. DUBs possess conserved catalytic regions known as the 'cys domain' and the 'his domain.' There are 2 distinct families of DUBs: ubiquitin-specific processing proteases (UBPs, or USPs) and ubiquitin C-terminal hydrolases (UCHs).
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP1
Applications Western Blot
Classification Polyclonal
Concentration 2.77 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene USP1
Gene Accession No. O94782
Gene Alias Deubiquitinating enzyme 1; EMBL:AAH92574.1, ECO:0000312; hUBP; MGC137005 protein; RGD:1306461}; ubiquitin carboxyl terminal hydrolase 1; ubiquitin carboxyl-terminal hydrolase 1; ubiquitin carboxyl-terminal hydrolase 1-like protein; ubiquitin specific peptidase 1; ubiquitin specific processing protease 1; ubiquitin specific protease 1; ubiquitin thioesterase 1; ubiquitin thiolesterase 1; ubiquitin-specific-processing protease 1; UBP; usp1; usp1 {ECO:0000312
Gene Symbols USP1
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human USP1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7398
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.