Learn More
USP1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301698AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
Deubiquitinating enzymes (DUBs) are capable of producing a free ubiquitin moiety from ubiquitin-fused precursors and ubiquitinylated proteins in cells. DUBs possess conserved catalytic regions known as the ′cys domain′ and the ′his domain.′ There are 2 distinct families of DUBs: ubiquitin-specific processing proteases (UBPs, or USPs) and ubiquitin C-terminal hydrolases (UCHs).Specifications
| USP1 | |
| Polyclonal | |
| Unconjugated | |
| USP1 | |
| Ubiquitin carboxyl-terminal hydrolase 1; Ubiquitin thioesterase 1; Ubiquitin-specific-processing protease 1; Deubiquitinating enzyme 1; hUBP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7398 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| O94782 | |
| USP1 | |
| Between 300 and 350 | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.