Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP10 Antibody (OTI2E1), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274798PCP
Description
USP10 Monoclonal specifically detects USP10 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry, Knockout Validated.Specifications
| USP10 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| USP10 | |
| Full length recombinant protein of human USP10 (NP_005144) produced in HEK293T cells. | |
| 0.1 mL | |
| 9100.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry, Immunoassay | |
| OTI2E1 | |
| Flow Cytometry, Knockout Validated | |
| Deubiquitinating enzyme 10, EC 3.4.19.12, KIAA0190MGC2621, ubiquitin carboxyl-terminal hydrolase 10, ubiquitin specific peptidase 10, ubiquitin specific protease 10, ubiquitin thioesterase 10, Ubiquitin thiolesterase 10, Ubiquitin-specific-processing protease 10, UBPO | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction