Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596706
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596706 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596706 Supplier Invitrogen™ Supplier No. PA596706
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FEPTYIYRLL TVNKSSLSEK GRQEDAEEYL GFILNGLHEE MLNLKKLLSP SNEKLTISNG PKNHSVNEEE QEEQGEGSED EWEQVGPRNK TSVTRQADFV QTPITGIFGG HIRSVVYQQS SKESATLQPF FTLQLDIQSD KIRTVQDALE SLVARESVQG YTTKTKQEVE ISRRVTLEKL PPVLVLHLKR FVYEKTGGCQ KLIKNIEYPV DLEISKELLS PGVKNKNFKC HRTYRLFAVV YHHGNSATGG HYTTDVFQIG LNGWLRIDDQ TVKVINQYQV VKPTAERTAY LLYYRRVDLL; Positive Samples: HepG2, Mouse brain; Cellular Location: Cytoplasm, Early endosome, Nucleus.

USP10, also known as ubiquitin specific peptidase 10, belongs to the ubiquitin-specific protease family of cysteine proteases. USP10 functions to catalyze the cleavage of ubiquitin from ubiquitin-conjugated protein substrates such as p53/TP53, SNX3 and CFTR. USP10 has been identified as a subunit of DNA-bound androgen receptor (AR) complexes and may play a role in the activity of the DNA-bound androgen receptor complex. USP10 also acts as an essential regulator of p53/TP53 stability and is thought to function as a tumor suppressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP10
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.12 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Usp10
Gene Accession No. P52479, Q14694
Gene Alias 2610014N07Rik; Deubiquitinating enzyme 10; KIAA0190; MGC2621; mKIAA0190; Ode-1; ubiquintin c-terminal hydrolase related polypeptide; ubiquitin carboxyl-terminal hydrolase 10; ubiquitin specific peptidase 10; ubiquitin specific protease 10; ubiquitin thioesterase 10; ubiquitin thiolesterase 10; ubiquitin-specific-processing protease 10; UBPO; Uchrp; USP10
Gene Symbols Usp10
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 499-798 of human USP10 (NP_0051442).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22224, 9100
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.