Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP11 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP18063925UL
Description
USP11 Polyclonal specifically detects USP11 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
USP11 | |
Polyclonal | |
Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
P51784, P51784, P51784, P51784 | |
USP11 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:VEPQEDTRLWAKNSEGSLDRLYDTHITVLDAALETGQLIIMETRKKDGTWPSAQLHVMNNNMSEEDEDFKGQPGICGLTNLGNT | |
25ul | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
Deubiquitinating enzyme 11, EC 3.1.2.15, EC 3.4.19.12, ubiquitin carboxyl-terminal hydrolase 11, ubiquitin specific peptidase 11, ubiquitin specific protease 11, ubiquitin thioesterase 11, Ubiquitin thiolesterase 11, ubiquitin-specific processing protease 11, Ubiquitin-specific-processing protease 11, UHX1ubiquitin carboxyl-terminal hydrolase, X-linked | |
Rabbit | |
Affinity Purified | |
RUO | |
8237 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction