Learn More
Invitrogen™ USP11 Polyclonal Antibody

Description
Recommended positive controls: HeLaS3, IMR32. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | USP11 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | USP11 |
| Gene Accession No. | P51784 |
| Gene Alias | 6230415D12Rik; deubiquitinating enzyme 11; mKIAA4085; RP4-659F15.2; Ubiquitin carboxyl-terminal hydrolase 11; ubiquitin carboxyl-terminal hydrolase, X-linked; ubiquitin specific peptidase 11; ubiquitin specific protease 11; ubiquitin thioesterase 11; ubiquitin thiolesterase 11; ubiquitin-specific processing protease 11; ubiquitin-specific-processing protease 11; UHX1; USP11 |
| Gene Symbols | USP11 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.