Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598233
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598233 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598233 Supplier Invitrogen™ Supplier No. PA598233
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

USP12 Polyclonal Antibody for Western Blot, ELISA

Deubiquitinating enzyme. Has almost no deubiquitinating activity by itself and requires the interaction with WDR20 and WDR48 to have a high activity (PubMed:19075014, PubMed:27373336). Not involved in deubiquitination of monoubiquitinated FANCD2 (PubMed:19075014). In complex with WDR48, acts as a potential tumor suppressor by positively regulating PHLPP1 stability (PubMed:24145035). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP12
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene USP12
Gene Accession No. O75317, Q9D9M2
Gene Alias Deubiquitinating enzyme 12; UBH1; Ubiquitin carboxyl-terminal hydrolase 12; ubiquitin hydrolyzing enzyme 1; ubiquitin specific peptidase 12; ubiquitin specific protease 12; ubiquitin specific protease 12 like 1; ubiquitin thioesterase 12; ubiquitin thiolesterase 12; ubiquitin-hydrolyzing enzyme 1; Ubiquitin-specific-processing protease 12; Usp12; USP12L1
Gene Symbols USP12
Host Species Rabbit
Immunogen Recombinant Human Ubiquitin carboxyl-terminal hydrolase 12 protein (1-370AA).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219333, 22217
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.