Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139844
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139844 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139844 Supplier Invitrogen™ Supplier No. PA5139844
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: RLPNGNIDNE NNNSTPDPTW VHEI Highest antigen sequence indentity to the following orthologs: Mouse - 83%, Rat - 79%.

Deubiquitinating enzyme. Has almost no deubiquitinating activity by itself and requires the interaction with WDR20 and WDR48 to have a high activity (PubMed:19075014, PubMed:27373336). Not involved in deubiquitination of monoubiquitinated FANCD2 (PubMed:19075014). In complex with WDR48, acts as a potential tumor suppressor by positively regulating PHLPP1 stability (PubMed:24145035). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP12
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene USP12
Gene Accession No. O75317
Gene Alias Deubiquitinating enzyme 12; UBH1; Ubiquitin carboxyl-terminal hydrolase 12; ubiquitin hydrolyzing enzyme 1; ubiquitin specific peptidase 12; ubiquitin specific protease 12; ubiquitin specific protease 12 like 1; ubiquitin thioesterase 12; ubiquitin thiolesterase 12; ubiquitin-hydrolyzing enzyme 1; Ubiquitin-specific-processing protease 12; Usp12; USP12L1
Gene Symbols USP12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human USP12. Recombinant protein control fragment (Product #RP-108970).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219333
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.