Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ USP12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110554
Description
Immunogen sequence: MEILMTVSKF ASICTMGANA SALEKEIGPE QFPVNEHYFG LVNFGNTCYC NSVLQALYFC RPFREKVLAY KSQPRKKESL LTCLADLFHS IATQKKKVGV.
Deubiquitinating enzyme. Has almost no deubiquitinating activity by itself and requires the interaction with WDR20 and WDR48 to have a high activity (PubMed:19075014, PubMed:27373336). Not involved in deubiquitination of monoubiquitinated FANCD2 (PubMed:19075014). In complex with WDR48, acts as a potential tumor suppressor by positively regulating PHLPP1 stability (PubMed:24145035). [UniProt].
Specifications
USP12 | |
Polyclonal | |
Unconjugated | |
USP12 | |
Deubiquitinating enzyme 12; UBH1; Ubiquitin carboxyl-terminal hydrolase 12; ubiquitin hydrolyzing enzyme 1; ubiquitin specific peptidase 12; ubiquitin specific protease 12; ubiquitin specific protease 12 like 1; ubiquitin thioesterase 12; ubiquitin thiolesterase 12; ubiquitin-hydrolyzing enzyme 1; Ubiquitin-specific-processing protease 12; Usp12; USP12L1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
219333, 22217, 360763 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.79 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
O75317, Q9D9M2 | |
USP12 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human USP12 (NP_8722942). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction