Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP13 Mouse anti-Canine, Human, Clone: OTI4G11, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA500318

Description
USP13, also known as Ubiquitin carboxyl-terminal hydrolase 13, is a deubiquitinating enzyme involved in ubiquitin-dependent protein catabolism. A member of the peptidase C19 family, USP13 is a highly conserved protein and contains two UBA domains and a UBP-type zinc finger. Expression of USP13 is higher in ovary and testis.Specifications
USP13 | |
Monoclonal | |
0.41 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q92995 | |
USP13 | |
Full-length protein expressed in 293T cell transfected with human USP13 expression vector | |
100 μL | |
Primary | |
Canine, Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
OTI4G11 | |
Unconjugated | |
USP13 | |
ISOT3, IsoT-3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
478640, 8975 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction