Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP13 Mouse anti-Human, Clone: 6E8C10, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | USP13 |
---|---|
Clone | 6E8C10 |
Concentration | 0.8 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6610
![]() ![]() |
Proteintech Group Inc
661761IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
USP13, also known as Ubiquitin carboxyl-terminal hydrolase 13, is a deubiquitinating enzyme involved in ubiquitin-dependent protein catabolism. A member of the peptidase C19 family, USP13 is a highly conserved protein and contains two UBA domains and a UBP-type zinc finger. Expression of USP13 is higher in ovary and testis.Specifications
USP13 | |
0.8 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Deubiquitinating enzyme 13, Isopeptidase T 3, IsoT 3, ISOT3, Ubiquitin thiolesterase 13, USP13 | |
USP13 | |
IgG1 | |
Protein G | |
Antibody |
6E8C10 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q92995 | |
8975 | |
USP13 Fusion Protein Ag10506 | |
Primary | |
-20°C | |
USP13 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title