Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP13 Mouse anti-Human, Clone: OTI5B9, liquid, TrueMAB™
SDP

Catalog No. 501671502
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-1502 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-1502 Supplier OriGene Supplier No. TA500367
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

USP13, also known as Ubiquitin carboxyl-terminal hydrolase 13, is a deubiquitinating enzyme involved in ubiquitin-dependent protein catabolism. A member of the peptidase C19 family, USP13 is a highly conserved protein and contains two UBA domains and a UBP-type zinc finger. Expression of USP13 is higher in ovary and testis.

Specifications

Antigen USP13
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone OTI5B9
Concentration 0.83 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene USP13
Gene Accession No. Q92995
Gene Alias ISOT3, IsoT-3
Gene Symbols USP13
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human USP13 expression vector
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8975
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.