Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP14 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP186946
Description
USP14 Polyclonal specifically detects USP14 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
USP14 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:500-1:1000 | |
P54578 | |
USP14 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:EKESVNAKVLKDVKFPLMLDMYELCTPELQEKMVSFRSKFKDLEDKKVNQQPNTSDKKSSPQKEVKYEPFSFADDIGSNNCGYYDLQAVLTHQGRSSSSGHYVSWVKRKQDEWIKFDDDKVSIVTPEDILRLSGGGDWHIAYVL | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
Deubiquitinating enzyme 14, EC 3.1.2.15, EC 3.4.19.12, TGT, TGTubiquitin carboxyl-terminal hydrolase 14, tRNA-guanine transglycosylase, 60-kD subunit, ubiquitin specific peptidase 14 (tRNA-guanine transglycosylase), ubiquitin specific protease 14 (tRNA-guanine transglycosylase), ubiquitin thioesterase 14, Ubiquitin thiolesterase 14, ubiquitin-specific processing protease 14, Ubiquitin-specific-processing protease 14 | |
Rabbit | |
56 kDa | |
0.1 mL | |
Protein Turnover, Ubiquitin Proteasome Pathway | |
9097 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction