Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530301
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530301 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530301 Supplier Invitrogen™ Supplier No. PA530301
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa, A431, mouse brain, rat brain. Predicted reactivity: Mouse (97%), Rat (97%), Xenopus laevis (88%), Rabbit (97%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a protein that is one of many involved in mechanisms to ensure proper chromosome segregation during cell division.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP14
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Usp14
Gene Accession No. P54578, Q9JMA1
Gene Alias 2610005K12Rik; 2610037B11Rik; ataxia; AW107924; ax; C78769; deubiquitinating enzyme 14; dUB-type TGT for deubiquitinating enzyme; nmf375; TGT; tRNA-guanine transglycosylase, 60-kD subunit; ubiquitin carboxyl-terminal hydrolase 14; ubiquitin specific peptidase 14; ubiquitin specific peptidase 14 (tRNA-guanine transglycosylase); ubiquitin specific protease 14; ubiquitin specific protease 14 (tRNA-guanine transglycosylase); ubiquitin thioesterase 14; ubiquitin thiolesterase 14; ubiquitin-specific processing protease 14; ubiquitin-specific-processing protease 14; Usp14; wu:fk63d09; zgc:55949
Gene Symbols Usp14
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human USP14. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 291796, 59025, 9097
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.