Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP14 Rabbit anti-Human, Mouse, Rat, Clone: 3A3J1, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP315796100UL
This item is not returnable.
View return policy
Description
USP14 Monoclonal antibody specifically detects USP14 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
USP14 | |
Monoclonal | |
Unconjugated | |
Deubiquitinating enzyme 14, EC 3.1.2.15, EC 3.4.19.12, TGT, TGTubiquitin carboxyl-terminal hydrolase 14, tRNA-guanine transglycosylase, 60-kD subunit, ubiquitin specific peptidase 14 (tRNA-guanine transglycosylase), ubiquitin specific protease 14 (tRNA-guanine transglycosylase), ubiquitin thioesterase 14, Ubiquitin thiolesterase 14, ubiquitin-specific processing protease 14, Ubiquitin-specific-processing protease 14 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 371-473 of human USP14 (P54578). | |
100 μg | |
Protein Turnover, Ubiquitin Proteasome Pathway | |
9097 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunoprecipitation | |
3A3J1 | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction