Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP14 Rabbit anti-Human, Mouse, Rat, Clone: 3A3J1, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP31579620UL
Description
USP14 Monoclonal antibody specifically detects USP14 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
USP14 | |
Monoclonal | |
Unconjugated | |
Deubiquitinating enzyme 14, EC 3.1.2.15, EC 3.4.19.12, TGT, TGTubiquitin carboxyl-terminal hydrolase 14, tRNA-guanine transglycosylase, 60-kD subunit, ubiquitin specific peptidase 14 (tRNA-guanine transglycosylase), ubiquitin specific protease 14 (tRNA-guanine transglycosylase), ubiquitin thioesterase 14, Ubiquitin thiolesterase 14, ubiquitin-specific processing protease 14, Ubiquitin-specific-processing protease 14 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 371-473 of human USP14 (P54578). | |
20 μg | |
Protein Turnover, Ubiquitin Proteasome Pathway | |
9097 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunoprecipitation | |
3A3J1 | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction