Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP15 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11040690R
Description
USP15 Polyclonal antibody specifically detects USP15 in Human, Mouse samples. It is validated for Western BlotSpecifications
USP15 | |
Polyclonal | |
Western Blot | |
Deubiquitinating enzyme 15, EC 3.1.2.15, EC 3.4.19.12, KIAA0529ubiquitin carboxyl-terminal hydrolase 15, MGC131982, MGC149838, MGC74854, ubiquitin specific peptidase 15, ubiquitin specific protease 15, ubiquitin thioesterase 15, Ubiquitin thiolesterase 15, Ubiquitin-specific-processing protease 15, unph-2, UNPH4 | |
The immunogen recognized by this antibody maps to a region between residue 960 and the C-terminus of human Ubiquitin Specific Peptidase 15 (AAD41086.1). | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
9958 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction