Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP15 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11040690
Description
USP15 Polyclonal antibody specifically detects USP15 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation, Knockdown.Specifications
USP15 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
Deubiquitinating enzyme 15, EC 3.1.2.15, EC 3.4.19.12, KIAA0529ubiquitin carboxyl-terminal hydrolase 15, MGC131982, MGC149838, MGC74854, ubiquitin specific peptidase 15, ubiquitin specific protease 15, ubiquitin thioesterase 15, Ubiquitin thiolesterase 15, Ubiquitin-specific-processing protease 15, unph-2, UNPH4 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Based on 100% sequence identity, this antibody is predicted to react with X. tropicalis, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Tasmanian devil, White-tufted-ear marmoset and Crab-eating macaque. | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation, KnockDown, Immunohistochemistry (Paraffin), KnockDown | |
1.0 mg/mL | |
Western Blot 1:2000 - 1:10000, Immunoprecipitation 2-10 ug/mg lysate, KnockDown Validated | |
Q9Y4E8 | |
USP15 | |
The immunogen recognized by this antibody maps to a region between residue 960 and the C-terminus of human Ubiquitin Specific Peptidase 15 (AAD41086.1). | |
100 μL | |
DNA replication Transcription Translation and Splicing | |
9958 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction