Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP15 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501572212
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-2212 10 μL
50-157-2211 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-2212 Supplier Bethyl Laboratories, Inc Supplier No. IHC00507T
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with X. tropicalis, Chicken, Turkey, Bovine, Dog, Horse, Rabbit, Pig, Panda, Orangutan, Monkey, Gorill

Ubiquitin (MIM 191339), a highly conserved protein involved in the regulation of intracellular protein breakdown, cell cycle regulation, and stress response, is released from degraded proteins by disassembly of the polyubiquitin chains. The disassembly process is mediated by ubiquitin-specific proteases (USPs). Also see USP1 (MIM 603478).

Specifications

Antigen USP15
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene USP15
Gene Accession No. Q8R5H1, Q9Y4E8
Gene Alias Ubiquitin carboxyl-terminal hydrolase 15; Ubiquitin thiolesterase 15; Ubiquitin-specific processing protease 15; Deubiquitinating enzyme 15; Unph-2; Unph4
Gene Symbols USP15
Host Species Rabbit
Immunogen Between 960 and C-term
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14479, 9958
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.