Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP16 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10081638JF646
Description
USP16 Polyclonal antibody specifically detects USP16 in Human samples. It is validated for Western BlotSpecifications
USP16 | |
Polyclonal | |
Western Blot | |
Deubiquitinating enzyme 16, EC 3.1.2.15, EC 3.4.19.12, human ubiquitin processing protease, EC 3.1.2.1510ubiquitin specific protease 16, ubiquitin carboxyl-terminal hydrolase 16, ubiquitin specific peptidase 16, ubiquitin thioesterase 16, Ubiquitin thiolesterase 16, Ubiquitin-processing protease UBP-M, ubiquitin-specific processing protease 16, Ubiquitin-specific-processing protease 16, UBP-M | |
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human ubiquitin specific peptidase 16 using the numbering given in entry NP_006438.1 (GeneID 10600). | |
0.1 mL | |
Cell Cycle and Replication | |
10600 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction