Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP16 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10081638
Description
USP16 Polyclonal specifically detects USP16 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).Specifications
USP16 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
Deubiquitinating enzyme 16, EC 3.1.2.15, EC 3.4.19.12, human ubiquitin processing protease, EC 3.1.2.1510ubiquitin specific protease 16, ubiquitin carboxyl-terminal hydrolase 16, ubiquitin specific peptidase 16, ubiquitin thioesterase 16, Ubiquitin thiolesterase 16, Ubiquitin-processing protease UBP-M, ubiquitin-specific processing protease 16, Ubiquitin-specific-processing protease 16, UBP-M | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Immunoprecipitation, Western Blot | |
1.0 mg/mL | |
Immunoprecipitation 1:10-1:500, Western Blot (Negative) | |
NP_006438.1 | |
USP16 | |
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human ubiquitin specific peptidase 16 using the numbering given in entry NP_006438.1 (GeneID 10600). | |
100 μL | |
Cell Cycle and Replication | |
10600 | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction