Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP16 Antibody (OTI1B4), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274805B
Description
USP16 Monoclonal specifically detects USP16 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot.Specifications
USP16 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
USP16 | |
Recombinant protein fragment corresponding to amino acids 137-466 of human USP16 (NP_001001992) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG2a |
Western Blot | |
OTI1B4 | |
Western Blot | |
Deubiquitinating enzyme 16, EC 3.1.2.15, EC 3.4.19.12, human ubiquitin processing protease, EC 3.1.2.1510ubiquitin specific protease 16, ubiquitin carboxyl-terminal hydrolase 16, ubiquitin specific peptidase 16, ubiquitin thioesterase 16, Ubiquitin thiolesterase 16, Ubiquitin-processing protease UBP-M, ubiquitin-specific processing protease 16, Ubiquitin-specific-processing protease 16, UBP-M | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication | |
10600.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction