Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP16 Monoclonal Antibody (OTI1B4)
GREENER_CHOICE

Catalog No. PIMA526205
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526205 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526205 Supplier Invitrogen™ Supplier No. MA526205
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

USP16 Monoclonal Antibody for Western Blot

Modification of target proteins by ubiquitin participates in a wide array of biological functions. Proteins destined for degradation or processing via the 26 S proteasome are coupled to multiple copies of ubiquitin. However, attachment of ubiquitin or ubiquitin-related molecules may also result in changes in subcellular distribution or modification of protein activity. An additional level of ubiquitin regulation, deubiquitination, is catalyzed by proteases called deubiquitinating enzymes, which fall into four distinct families. Ubiquitin C-terminal hydrolases, ubiquitin-specific processing proteases (USPs),1 OTU-domain ubiquitin-aldehyde-binding proteins, and Jab1/Pad1/MPN-domain-containing metallo-enzymes. Among these four families, USPs represent the most widespread and represented deubiquitinating enzymes across evolution. USPs tend to release ubiquitin from a conjugated protein. They display similar catalytic domains containing conserved Cys and His boxes but divergent N-terminal and occasionally C-terminal extensions, which are thought to function in substrate recognition, subcellular localization, and protein-protein interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP16
Applications Western Blot
Classification Monoclonal
Clone OTI1B4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene USP16
Gene Accession No. Q9Y5T5
Gene Alias 1200004E02Rik; 2810483I07Rik; 6330514E22Rik; Deubiquitinating enzyme 16; deubiquitinating enzyme 16 {ECO:0000255; HAMAP-Rule:MF_03062}; MSTP039; Ubiquitin carboxyl-terminal hydrolase 16; ubiquitin carboxyl-terminal hydrolase 16 {ECO:0000255; Ubiquitin carboxyl-terminal hydrolase 16-like protein; ubiquitin specific peptidase 16; ubiquitin specific protease 16; ubiquitin thioesterase 16; ubiquitin thioesterase 16 {ECO:0000255; ubiquitin thiolesterase 16; ubiquitin-processing protease UBP-M; ubiquitin-specific processing protease 16; ubiquitin-specific-processing protease 16; ubiquitin-specific-processing protease 16 {ECO:0000255; UBPM; UBP-M; USP16
Gene Symbols USP16
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 137-466 of USP16 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10600
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.