Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP16 Polyclonal Antibody
USP16 Polyclonal Antibody
Supplier: Thermo Scientific PA542775
Description
USP16 Polyclonal Antibody for Western Blot, ICC/IF, IHC

Specifications
USP16 | |
Polyclonal | |
Unconjugated | |
USP16 | |
1200004E02Rik; 2810483I07Rik; 6330514E22Rik; Deubiquitinating enzyme 16; deubiquitinating enzyme 16 {ECO:0000255; HAMAP-Rule:MF_03062}; MSTP039; Ubiquitin carboxyl-terminal hydrolase 16; ubiquitin carboxyl-terminal hydrolase 16 {ECO:0000255; Ubiquitin carboxyl-terminal hydrolase 16-like protein; ubiquitin specific peptidase 16; ubiquitin specific protease 16; ubiquitin thioesterase 16; ubiquitin thioesterase 16 {ECO:0000255; ubiquitin thiolesterase 16; ubiquitin-processing protease UBP-M; ubiquitin-specific processing protease 16; ubiquitin-specific-processing protease 16; ubiquitin-specific-processing protease 16 {ECO:0000255; UBPM; UBP-M; USP16 | |
Rabbit | |
Protein A | |
RUO | |
10600 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9Y5T5 | |
USP16 | |
Synthetic peptide directed towards the N-terminal of human USP16 (aa 51-100). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction