Learn More
Invitrogen™ USP16 Polyclonal Antibody

Description
Immunogen sequence: CLVLSLDNWS VWCYVCDNEV QYCSSNQLGQ VVDYVRKQAS ITTPKPAEKD NGNIELENKK LEKESKNEQE REKKENMAKE NPPMNSPCQI TVKGLSNLGN TCFFNAVMQN LSQTPVLREL LKEVKM Highest antigen sequence indentity to the following orthologs: Rat - 84%, Mouse - 86%.
Specifications
Specifications
| Antigen | USP16 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | USP16 |
| Gene Accession No. | Q9Y5T5 |
| Gene Alias | 1200004E02Rik; 2810483I07Rik; 6330514E22Rik; Deubiquitinating enzyme 16; deubiquitinating enzyme 16 {ECO:0000255; HAMAP-Rule:MF_03062}; MSTP039; Ubiquitin carboxyl-terminal hydrolase 16; ubiquitin carboxyl-terminal hydrolase 16 {ECO:0000255; Ubiquitin carboxyl-terminal hydrolase 16-like protein; ubiquitin specific peptidase 16; ubiquitin specific protease 16; ubiquitin thioesterase 16; ubiquitin thioesterase 16 {ECO:0000255; ubiquitin thiolesterase 16; ubiquitin-processing protease UBP-M; ubiquitin-specific processing protease 16; ubiquitin-specific-processing protease 16; ubiquitin-specific-processing protease 16 {ECO:0000255; UBPM; UBP-M; USP16 |
| Gene Symbols | USP16 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.