Learn More
Invitrogen™ USP16 Polyclonal Antibody

Description
Antibody detects endogenous levels of total USP16.
Specifications
Specifications
| Antigen | USP16 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | USP16 |
| Gene Accession No. | Q2KJ09, Q99LG0, Q9Y5T5 |
| Gene Alias | 1200004E02Rik; 2810483I07Rik; 6330514E22Rik; Deubiquitinating enzyme 16; deubiquitinating enzyme 16 {ECO:0000255; HAMAP-Rule:MF_03062}; MSTP039; Ubiquitin carboxyl-terminal hydrolase 16; ubiquitin carboxyl-terminal hydrolase 16 {ECO:0000255; Ubiquitin carboxyl-terminal hydrolase 16-like protein; ubiquitin specific peptidase 16; ubiquitin specific protease 16; ubiquitin thioesterase 16; ubiquitin thioesterase 16 {ECO:0000255; ubiquitin thiolesterase 16; ubiquitin-processing protease UBP-M; ubiquitin-specific processing protease 16; ubiquitin-specific-processing protease 16; ubiquitin-specific-processing protease 16 {ECO:0000255; UBPM; UBP-M; USP16 |
| Gene Symbols | USP16 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.