Learn More
USP16 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301615AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine
This gene encodes a deubiquitinating enzyme that is phosphorylated at the onset of mitosis and then dephosphorylated at the metaphase/anaphase transition. It can deubiquitinate H2A, one of two major ubiquitinated proteins of chromatin, in vitro and a mutant form of the protein was shown to block cell division. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.Specifications
| USP16 | |
| Polyclonal | |
| Unconjugated | |
| USP16 | |
| Ubiquitin thiolesterase 16; Ubiquitin-specific processing protease 16; Deubiquitinating enzyme 16; Ubiquitin processing protease UBP-M | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10600 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 1 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q9Y5T5 | |
| USP16 | |
| Between 225 and 275 | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.