Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP17L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568353
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568353 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568353 Supplier Invitrogen™ Supplier No. PA568353
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse and Rat samples.

USP17L111 is a deubiquitinating enzyme that removes conjugated ubiquitin from specific proteins to regulate different cellular processes that may include cell proliferation, progression through the cell cycle, apoptosis, cell migration, and the cellular response to viral infection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP17L1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene USP17L1
Gene Accession No. Q7RTZ2
Gene Alias deubiquitinating enzyme 17-like protein 1; putative ubiquitin carboxyl-terminal hydrolase 17-like protein 1; ubiquitin carboxyl-terminal hydrolase 17-like protein 1; ubiquitin specific peptidase 17-like family member 1; ubiquitin thioesterase 17-like protein 1; Ubiquitin-specific-processing protease 17-like protein 1; USP17L; USP17L1; USP17L1P
Gene Symbols USP17L1
Host Species Rabbit
Immunogen Synthesized peptide derived from human USP17L1.
Purification Method affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 401447
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.