Learn More
Invitrogen™ USP17L2 Polyclonal Antibody

Description
USP17L2 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | USP17L2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | USP17L2 |
| Gene Accession No. | Q6R6M4 |
| Gene Alias | deubiquitinating enzyme 17-like protein 2; deubiquitinating enzyme 3; deubiquitinating protein 3; DUB3; DUB-3; Ubiquitin carboxyl-terminal hydrolase 17; ubiquitin carboxyl-terminal hydrolase 17-like protein 2; ubiquitin specific peptidase 17-like 2; ubiquitin specific peptidase 17-like family member 2; ubiquitin thioesterase 17-like protein 2; ubiquitin thiolesterase 17-like protein 2; ubiquitin-specific-processing protease 17-like protein 2; USP17; USP17H; USP17I; USP17J; USP17K; USP17L; USP17L2; USP17M |
| Gene Symbols | USP17L2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.