Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP17L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544961
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544961 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544961 Supplier Invitrogen™ Supplier No. PA544961
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: FYIQKSEWER HSESVSRGRE PRALGAEDTD RRATQGELKR DHPCLQAPEL Sequence homology: Human: 100%.

DUB3 is a member of the ubiquitin processing protease (UBP) subfamily of deubiquitinating enzymes. See USP1 (MIM 603478) for background information.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP17L2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene USP17L2
Gene Accession No. Q6R6M4
Gene Alias deubiquitinating enzyme 17-like protein 2; deubiquitinating enzyme 3; deubiquitinating protein 3; DUB3; DUB-3; Ubiquitin carboxyl-terminal hydrolase 17; ubiquitin carboxyl-terminal hydrolase 17-like protein 2; ubiquitin specific peptidase 17-like 2; ubiquitin specific peptidase 17-like family member 2; ubiquitin thioesterase 17-like protein 2; ubiquitin thiolesterase 17-like protein 2; ubiquitin-specific-processing protease 17-like protein 2; USP17; USP17H; USP17I; USP17J; USP17K; USP17L; USP17L2; USP17M
Gene Symbols USP17L2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human USP17L2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 377630
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.