Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP17L3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112880
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112880 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112880 Supplier Thermo Scientific Supplier No. PA5112880
Add to Cart
Add to Cart

USP17L3 Polyclonal Antibody for Western Blot, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen USP17L3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene USP17L3
Gene Accession No. A6NCW0
Gene Alias Deubiquitinating enzyme 17-like protein 3; ubiquitin carboxyl-terminal hydrolase 17-like protein 3; ubiquitin specific peptidase 17-like 3; ubiquitin specific peptidase 17-like family member 3; ubiquitin thioesterase 17-like protein 3; Ubiquitin-specific-processing protease 17-like protein 3; USP17B; USP17F; USP17L3
Gene Symbols USP17L3
Host Species Rabbit
Immunogen Recombinant Human Ubiquitin carboxyl-terminal hydrolase 17-like protein 3 protein (19-251AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 645836
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.