Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ USP17L3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546659
Description
Peptide sequence: SESVSRGREP RALGAEDTDR RAKQGELKRD HPCLQAPELD EHLVERATQE Sequence homology: Human: 100%.
USP17L3 is a deubiquitinating enzyme that removes conjugated ubiquitin from specific proteins to regulate different cellular processes that may include cell proliferation, progression through the cell cycle, apoptosis, cell migration, and the cellular response to viral infection.
Specifications
USP17L3 | |
Polyclonal | |
Unconjugated | |
USP17L3 | |
Deubiquitinating enzyme 17-like protein 3; ubiquitin carboxyl-terminal hydrolase 17-like protein 3; ubiquitin specific peptidase 17-like 3; ubiquitin specific peptidase 17-like family member 3; ubiquitin thioesterase 17-like protein 3; Ubiquitin-specific-processing protease 17-like protein 3; USP17B; USP17F; USP17L3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
645836 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
A6NCW0 | |
USP17L3 | |
Synthetic peptide directed towards the C-terminal of human USP17L3 (aa 382-431). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction