Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP17L7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546660
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546660 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546660 Supplier Invitrogen™ Supplier No. PA546660
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KNHHPEQQSS LLNLSSTKPT DQESMNTGTL ASLQGSTRRS KGNNKHSKRS.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP17L7
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene USP17L7
Gene Accession No. P0C7H9
Gene Alias Inactive ubiquitin carboxyl-terminal hydrolase 17-like protein 7; ubiquitin specific peptidase 17-like 7; ubiquitin specific peptidase 17-like family member 7; USP17L7
Gene Symbols USP17L7
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human USP17L7 (aa 476-525).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 392197
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.