Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102960
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102960 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102960 Supplier Invitrogen™ Supplier No. PA5102960
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total USP18.

USP18, a member of the deubiquitinating protease family of enzymes, removes ubiquitin adducts from a broad range of protein substrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP18
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene USP18
Gene Accession No. Q9UMW8
Gene Alias 1110058H21Rik; 43 kDa ISG15-specific protease; AW047653; bubp43; hUBP43; ISG15-specific-processing protease; ISG43; putative ubiquitin-specific protease; ubiquitin specific peptidase 18; ubiquitin specific protease 15; ubiquitin specific protease 18; ubl carboxyl-terminal hydrolase 18; Ubl thioesterase 18; ubl thiolesterase 18; Ubp15; UBP43; USP18
Gene Symbols USP18
Host Species Rabbit
Immunogen A synthesized peptide derived from human USP18(Accession Q9UMW8), corresponding to amino acid residues L16-L66.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11274
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.