Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542843
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542843 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542843 Supplier Invitrogen™ Supplier No. PA542843
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MQDSRQKAVR PLELAYCLQK CNVPLFVQHD AAQLYLKLWN LIKDQITDVH Sequence homology: Cow: 79%; Dog: 86%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 85%; Rabbit: 92%; Rat: 85%.

USP18, a member of the deubiquitinating protease family of enzymes, removes ubiquitin adducts from a broad range of protein substrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP18
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene USP18
Gene Accession No. Q9UMW8, Q9WTV6
Gene Alias 1110058H21Rik; 43 kDa ISG15-specific protease; AW047653; bubp43; hUBP43; ISG15-specific-processing protease; ISG43; putative ubiquitin-specific protease; ubiquitin specific peptidase 18; ubiquitin specific protease 15; ubiquitin specific protease 18; ubl carboxyl-terminal hydrolase 18; Ubl thioesterase 18; ubl thiolesterase 18; Ubp15; UBP43; USP18
Gene Symbols USP18
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human USP18 (aa 110-159).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11274, 24110
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.