Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP19 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10079828B
Description
USP19 Polyclonal antibody specifically detects USP19 in Human samples. It is validated for Western BlotSpecifications
USP19 | |
Polyclonal | |
Western Blot | |
Deubiquitinating enzyme 19, EC 3.1.2.15, EC 3.4.19.12, KIAA0891, ubiquitin carboxyl-terminal hydrolase 19, ubiquitin specific peptidase 19, ubiquitin thioesterase 19, Ubiquitin thiolesterase 19, Ubiquitin-specific-processing protease 19, Zinc finger MYND domain-containing protein 9, ZMYND9ubiquitin specific protease 19 | |
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human ubiquitin specific peptidase 19 (zinc finger MYND domain-containing protein 9) using the numbering given in entry NP_006668.1 (GeneID 10869). | |
0.1 mL | |
Neuroscience | |
10869 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Biotin | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction