Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597239
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597239 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597239 Supplier Invitrogen™ Supplier No. PA597239
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSGGASATGP RRGPPGLEDT TSKKKQKDRA NQESKDGDPR KETGSRYVAQ AGLEPLASGD PSASASHAAG ITGSRHRTRL FFPSSSGSAS TPQEEQTKEG ACEDPHDLLA TPTPELLLDW RQSAEEVIVK LRVGVGPLQL EDVDAAFTDT DCVVRFAGGQ QWGGVFYAEI KSSCAKVQTR KGSLLHLTLP KKVPMLTWPS; Positive Samples: U-87MG, A-431, HeLa; Cellular Location: Endoplasmic reticulum membrane, Single-pass membrane protein.

Modification of target proteins by ubiquitin participates in a wide array of biological functions. Proteins destined for degradation or processing via the 26 S proteasome are coupled to multiple copies of ubiquitin. However, attachment of ubiquitin or ubiquitin-related molecules may also result in changes in subcellular distribution or modification of protein activity. An additional level of ubiquitin regulation, deubiquitination, is catalyzed by proteases called deubiquitinating enzymes, which fall into four distinct families. Ubiquitin C-terminal hydrolases, ubiquitin-specific processing proteases (USPs), 1 OTU-domain ubiquitin-aldehyde-binding proteins, and Jab1/Pad1/MPN-domain-containing metallo-enzymes. Among these four families, USPs represent the most widespread and represented deubiquitinating enzymes across evolution. USPs tend to release ubiquitin from a conjugated protein. They display similar catalytic domains containing conserved Cys and His boxes but divergent N-terminal and occasionally C-terminal extensions, which are thought to function in substrate recognition, subcellular localization, and protein-protein interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP19
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene USP19
Gene Accession No. O94966, Q3UJD6
Gene Alias 8430421I07Rik; AI047774; Deubiquitinating enzyme 19; KIAA0891; Ubiquitin carboxyl-terminal hydrolase 19; ubiquitin specific peptidase 19; ubiquitin specific protease 19; ubiquitin thioesterase 19; ubiquitin thiolesterase 19; ubiquitin-specific processing protease 19; ubiquitin-specific-processing protease 19; Usp19; Zinc finger MYND domain-containing protein 9; Zmynd9
Gene Symbols USP19
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human USP19 (NP_0011860891).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10869, 71472
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.