Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP2 Monoclonal Antibody (1738CT331.50.87)
GREENER_CHOICE

Catalog No. PIMA537724
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537724 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537724 Supplier Invitrogen™ Supplier No. MA537724
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

USP2 Monoclonal Antibody for Western Blot

Ubiquitin is a highly conserved protein involved in the regulation of intracellular protein breakdown, cell cycle regulation, and stress response, is released from degraded proteins by disassembly of the polyubiquitin chains. The disassembly process is mediated by ubiquitin-specific proteases (USPs).
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP2
Applications Western Blot
Classification Monoclonal
Clone 1738CT331.50.87
Concentration 0.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene USP2
Gene Accession No. O75604
Gene Alias 41 kDa ubiquitin-specific protease; B930035K21Rik; deubiquitinating enzyme 2; MNCb-0190; Ubiquitin carboxyl-terminal hydrolase 2; ubiquitin specific peptidase 2; ubiquitin specific protease 12; ubiquitin specific protease 2; ubiquitin specific protease 9; ubiquitin thioesterase 2; ubiquitin thiolesterase 2; ubiquitin-specific-processing protease 2; Ubiquitin-specific-processing protease testis; Ubp41; Ubp69; UBP-t; USP2; USP9
Gene Symbols USP2
Host Species Mouse
Immunogen recombinant protein between 1-258 amino acids from human USP2.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9099
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.